Learn More
Research Products International Corp Acrylic Dispensing Bin, with Magnetic Door, Medium, 6.25 x 10.5 x 9.5 Inches
SDP
Catalog No. 50998803
Encompass
Acrylic Dispensing Bins are convenient for storing and dispensing pipette tips, micro-tubes, stoppers, or other small laboratory items. Unique magnetic door allows quick access of contents. Dispensing Bins are constructed of heavy duty acrylic for rugged everyday use. Manufactured of 1/8" clear acrylic that allows full view of contents and withstands temperature down to -70°C.
Catalog No. 50-998-803 Supplier Research Products International Corp Supplier No. 246717
Acrylic Dispensing Bins are convenient for storing and dispensing pipette tips, micro-tubes, stoppers, or other small laboratory items. Unique magnetic door allows quick access of contents. Dispensing Bins are constructed of heavy duty acrylic for rugged everyday use. Manufactured of 1/8" clear acrylic that allows full view of contents and withstands temperature down to -70°C.