Learn More
Electron Microscopy Sciences Dispensing Bottle 150 ml, 31 mm diameter, 115 mm height
SDP
Catalog No. 5025504
Encompass_Preferred
Low density polyethylene bottles. Tip has a 0.003" (0.76 mm) opening. By squeezing the bottle a steady stream of liquid is produced.
Diameter: 31 mm
Height: 115 mm
Catalog No. 50-255-04 Supplier Electron Microscopy Sciences Supplier No. 64294
Low density polyethylene bottles. Tip has a 0.003" (0.76 mm) opening. By squeezing the bottle a steady stream of liquid is produced.
Diameter: 31 mm
Height: 115 mm