Learn More
BURKLE INC Dropper bottle Drop Boy, HDPE, 0.85 oz. (25 ml), with separate cap
Supplier:  BURKLE INC 03070025
Encompass_Preferred
The Drop-Boy is a dropper bottle for precision applications. Manufactured from tough, chemically resistant HDPE, it is ideal for liquids that need to be dispensed accurately in small quantities, drop by drop or as a splash.
The Drop-Boy dropper bottle provides precise drop-by-drop dosing for controlled and consistent dispensing of substances, minimizing waste and saving costs, especially with expensive contents.
In addition to accurate dosing, dropper bottles prevent contamination of the contents by allowing the user to dispense the liquid without direct contact.
The dropper bottle can be re-closed with a separate screw cap. The long and very pointed dropper tip is sealed and cut off before first use. The dropper tip can be shortened to adjust the drop size.
Dropper bottles are easy to use and are an effective solution for precise and hygienic administration of liquids.
Catalog No. 50-000-25615