Learn More
Electron Microscopy Sciences Dispensing Bottle 250 ml, 39 mm diameter, 128 mm height
SDP
Catalog No. 5025503
Encompass_Preferred
Low density polyethylene bottles. Tip has a 0.003" (0.76 mm) opening. By squeezing the bottle a steady stream of liquid is produced.
Diameter: 39 mm
Height: 128 mm
Catalog No. 50-255-03 Supplier Electron Microscopy Sciences Supplier No. 64295
Low density polyethylene bottles. Tip has a 0.003" (0.76 mm) opening. By squeezing the bottle a steady stream of liquid is produced.
Diameter: 39 mm
Height: 128 mm