Learn More
Electron Microscopy Sciences Dispensing Bottle 60 ml, 74 mm diameter, 196 mm height
SDP
Catalog No. 5025501
Encompass_Preferred
Low density polyethylene bottles. Tip has a 0.003" (0.76 mm) opening. By squeezing the bottle a steady stream of liquid is produced.
Diameter: 74 mm
Height: 196 mm
Catalog No. 50-255-01 Supplier Electron Microscopy Sciences Supplier No. 64292
Low density polyethylene bottles. Tip has a 0.003" (0.76 mm) opening. By squeezing the bottle a steady stream of liquid is produced.
Diameter: 74 mm
Height: 196 mm