Learn More
PATHOLOGY INNOVATIONS These 8 transparent vinyl slides provide an easy way to precisely place tissues into the embedding wells. The 8 slides allow placement in any orientation to the desired site in the well.
Catalog No. NC3370401
Encompass_Preferred
These thin transparent vinyl slides serve as a surface to precisely orient the tissue into the desired position and as a means to accurately transfer tissue to the embedding well floor using Peters Precision Cryoembedding System Well Bars. Tissue is applied face down to the end of the transparent slide where it can be visualized from below and manipulated into position. The face that is visible will be laid onto the well floor and will ultimately be the embedded surface to be sectioned. These dispensing slides were designed by professor and author of frozen section literature Dr. Stephen Peters.
Catalog No. NC3370401 Supplier PATHOLOGY INNOVATIONS Supplier No. 143
These thin transparent vinyl slides serve as a surface to precisely orient the tissue into the desired position and as a means to accurately transfer tissue to the embedding well floor using Peters Precision Cryoembedding System Well Bars. Tissue is applied face down to the end of the transparent slide where it can be visualized from below and manipulated into position. The face that is visible will be laid onto the well floor and will ultimately be the embedded surface to be sectioned. These dispensing slides were designed by professor and author of frozen section literature Dr. Stephen Peters.