Learn More
Med Vet International Sterile Small Volume 15mL Dispenser, J1209BS, 12 Count
Catalog No. 5000495766
Encompass_Preferred
These small volume dropper bottles are perfect for smaller pets or medications such as opthalmics. Also available in sterile packs• Bottle and cap made of slightly opaque, low density polyethylene• White cap made of polystyrene • Small drop volume inner nozzleSterile. 2 bottles per ETO sterilized pouch, 6 pouches per bag for a total of 12.15ml (0.5oz) sterile
Catalog No. 50-004-95766 Supplier Med Vet International Supplier No. JOR1209BS
These small volume dropper bottles are perfect for smaller pets or medications such as opthalmics. Also available in sterile packs• Bottle and cap made of slightly opaque, low density polyethylene• White cap made of polystyrene • Small drop volume inner nozzleSterile. 2 bottles per ETO sterilized pouch, 6 pouches per bag for a total of 12.15ml (0.5oz) sterile