Learn More
Med Vet International Sterile Small Volume 30mL Dispenser, 12/bag, J1209CS
Catalog No. 5000495603
Encompass_Preferred
These small volume dropper bottles are perfect for smaller pets or medications such as opthalmics. Also available in sterile packs• Bottle and cap made of slightly opaque, low density polyethylene• White cap made of polystyrene • Small drop volume inner nozzleSterile. 2 bottles per ETO sterilized pouch, 6 pouches per bag
Catalog No. 50-004-95603 Supplier Med Vet International Supplier No. JOR1209CS
These small volume dropper bottles are perfect for smaller pets or medications such as opthalmics. Also available in sterile packs• Bottle and cap made of slightly opaque, low density polyethylene• White cap made of polystyrene • Small drop volume inner nozzleSterile. 2 bottles per ETO sterilized pouch, 6 pouches per bag