Learn More
Jensen Global Inc flexible Polypro dispensing tips, Gauge: 18, Color: Green, 25/pk
Catalog No. NC2882660
Encompass_Preferred
Our flexible Polypro dispensing tips are manufactured with white polypropylene tubing. Best used in applications that need scratch-free results or the ability to insert the dispensing tip into a curved or problematic area. They can be cut to a customized length or to remove parts of the tip that have become difficult to use due to curing. Excellent when used with many adhesives including cyanoacrylates, anaerobics, or other quick curing adhesives. Available in boxes of 50 or bags of 1000.Gauge: 18 Color: GreenID: 0.033" (0.840 mm)OD: 0.053" (1.350 mm)
Catalog No. NC2882660 Supplier Jensen Global Inc Supplier No. JG180.5XPRO
Our flexible Polypro dispensing tips are manufactured with white polypropylene tubing. Best used in applications that need scratch-free results or the ability to insert the dispensing tip into a curved or problematic area. They can be cut to a customized length or to remove parts of the tip that have become difficult to use due to curing. Excellent when used with many adhesives including cyanoacrylates, anaerobics, or other quick curing adhesives. Available in boxes of 50 or bags of 1000.Gauge: 18 Color: GreenID: 0.033" (0.840 mm)OD: 0.053" (1.350 mm)