Learn More
Instech Replacement dispensing tip for Instech ABS2 automated blood sampler, 18ga.
SDP
Catalog No. 5080983
Encompass
The Instech ABS2/ND18 is an 18ga replacement dispensing tip designed for the ABS2 automated blood sampler, specifically for large‑animal tube set configurations. Its conductive 18‑gauge stainless design ensures smooth fluid delivery and accurate droplet formation during automated sampling cycles, supporting reliable performance in high‑flow workflows.
Features
  • 18ga dispensing tip for large‑animal ABS2 tube sets
  • Conductive stainless design for stable sampling
  • Provides accurate droplet formation and flow control
  • Direct replacement component for ABS2 systems
  • Supports high‑volume automated blood sampling
Catalog No. 50-809-83 Supplier Instech Supplier No. ABS2/ND18
The Instech ABS2/ND18 is an 18ga replacement dispensing tip designed for the ABS2 automated blood sampler, specifically for large‑animal tube set configurations. Its conductive 18‑gauge stainless design ensures smooth fluid delivery and accurate droplet formation during automated sampling cycles, supporting reliable performance in high‑flow workflows.
Features
  • 18ga dispensing tip for large‑animal ABS2 tube sets
  • Conductive stainless design for stable sampling
  • Provides accurate droplet formation and flow control
  • Direct replacement component for ABS2 systems
  • Supports high‑volume automated blood sampling