Learn More
Electron Microscopy Sciences Dispensing Trays, 16 x 20 x 4"
SDP
Catalog No. 5029509
Encompass_Preferred
These trays, which are 1/4" thick, are fabricated from either low or high density polyethylene. They come complete with a polyethylene dispensing valve which is located as close to the bottom as possible on the corner. The tray is suitable for specimen preparation procedures.
Size: 16 x 20 x 4"
Catalog No. 50-295-09 Supplier Electron Microscopy Sciences Supplier No. 6300702
These trays, which are 1/4" thick, are fabricated from either low or high density polyethylene. They come complete with a polyethylene dispensing valve which is located as close to the bottom as possible on the corner. The tray is suitable for specimen preparation procedures.
Size: 16 x 20 x 4"