Learn More
Electron Microscopy Sciences Dispensing Trays, 22 x 26 x 4"
SDP
Catalog No. 5029512
Encompass_Preferred
These trays, which are 1/4" thick, are fabricated from either low or high density polyethylene. They come complete with a polyethylene dispensing valve which is located as close to the bottom as possible on the corner. The tray is suitable for specimen preparation procedures.
Size: 22 x 26 x 4"
Catalog No. 50-295-12 Supplier Electron Microscopy Sciences Supplier No. 6300704
These trays, which are 1/4" thick, are fabricated from either low or high density polyethylene. They come complete with a polyethylene dispensing valve which is located as close to the bottom as possible on the corner. The tray is suitable for specimen preparation procedures.
Size: 22 x 26 x 4"