Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Disperse blue 1 | 2475-45-8 | MFCD00001225 | 30.0% | 268.27 g/mol | C14H12N4O2 | 50 MG
SDP

Catalog No. 5000281691
Encompass_Preferred

Disperse Blue 1 is an organic dye used in biological research to stain cells and tissues. It is supplied as a small-quantity powder for laboratory staining and analytical applications and is intended for research use only.

  • Organic dye used for staining cells and tissues.
  • Suitable for staining and microscopy applications.
  • Molecular formula C14H12N4O2; molecular weight 268.27 g/mol.
  • CAS number 2475-45-8 for substance identification.
  • Available as a 50 MG pack for small-scale experiments.
  • Sold for research use only.

Catalog No. 50-002-81691 Supplier Medchemexpress LLC Supplier No. HYW41444750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disperse Blue 1 is an organic dye used in biological research to stain cells and tissues. It is supplied as a small-quantity powder for laboratory staining and analytical applications and is intended for research use only.

  • Organic dye used for staining cells and tissues.
  • Suitable for staining and microscopy applications.
  • Molecular formula C14H12N4O2; molecular weight 268.27 g/mol.
  • CAS number 2475-45-8 for substance identification.
  • Available as a 50 MG pack for small-scale experiments.
  • Sold for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.