Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disperse blue 1, MP Biomedicals™

Catalog No. ICN15787505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15787505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15787505 Supplier MP Biomedicals, Inc Supplier No. 0215787505

Chemical Identifiers

CAS 2475-45-8
Molecular Formula C14H12N4O2
Molecular Weight (g/mol) 268.28
MDL Number MFCD00001225
InChI Key JSFUMBWFPQSADC-UHFFFAOYSA-N
Synonym 1,4,5,8-Tetraaminoanthraquinone, C.I. 64500
PubChem CID 17190
ChEBI CHEBI:82325
IUPAC Name 1,4,5,8-tetraamino-9,10-dihydroanthracene-9,10-dione
SMILES NC1=CC=C(N)C2=C1C(=O)C1=C(N)C=CC(N)=C1C2=O

Specifications

Formula Weight 268.3
Color Brown
Quantity 5 g
Physical Form Powder
Chemical Name or Material Disperse blue 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.