Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disperse Diazo Black 3BF, TCI America™
SDP

Catalog No. D161525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D161525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D161525G Supplier TCI America Supplier No. D161525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6232-57-1
Molecular Formula C14H16N4O
Molecular Weight (g/mol) 256.309
MDL Number MFCD00059857
InChI Key DBENZRUFCCLWPP-UHFFFAOYSA-N
PubChem CID 80382
IUPAC Name 4-[(4-aminophenyl)diazenyl]-2-methoxy-5-methylaniline
SMILES CC1=CC(=C(C=C1N=NC2=CC=C(C=C2)N)OC)N

Specifications

Quantity 25 g
Formula Weight 256.31
Physical Form Crystalline Powder
Chemical Name or Material Disperse Diazo Black 3BF
EINECSNumber (5)-2681
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.