Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Disperse Orange 1 | 2581-69-3 | 96.6% | 1 MG
SDP

Catalog No. 5000410254
Encompass_Preferred

Disperse Orange 1 is an orange-orange dye primarily used for fiber dyeing. It is intended for research use only and is not sold to patients. This product appears as a solid with a color ranging from purple to purplish red.

  • High purity: 96.60%
  • Solid appearance
  • Purple to purplish red color
  • Molecular weight: 318.33
  • Soluble in DMSO at 35 mg/mL
  • Stable when stored at 4°C, protected from light
  • Extended stability in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-10254 Supplier Medchemexpress LLC Supplier No. HYD03321MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disperse Orange 1 is an orange-orange dye primarily used for fiber dyeing. It is intended for research use only and is not sold to patients. This product appears as a solid with a color ranging from purple to purplish red.

  • High purity: 96.60%
  • Solid appearance
  • Purple to purplish red color
  • Molecular weight: 318.33
  • Soluble in DMSO at 35 mg/mL
  • Stable when stored at 4°C, protected from light
  • Extended stability in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.