Learn More
Medchemexpress LLC Disperse orange 1 | 2581-69-3 | MFCD00007311 | 96.6% | 318.33 | C18H14N4O2 | 10 MG
SDP
Catalog No. 5000297517
Encompass_Preferred
Disperse Orange 1 is an orange azo disperse dye (C18H14N4O2) used primarily for dyeing synthetic fibers such as polyester and nylon. Supplied in small laboratory pack sizes for research and analytical use, it is commonly used in textile dyeing studies and fluorescence applications. Accompanying datasheet and safety data sheet (SDS) provide handling and storage information.
  • High purity (approximately 96.6%) for analytical applications.
  • Molecular weight 318.33 for formulation calculations.
  • Suitable for dyeing polyester, nylon, and acrylic fibers.
  • Available in small laboratory pack sizes, including 10 mg, for method development.
  • Provided with a datasheet and SDS for safe handling and storage.
Catalog No. 50-002-97517 Supplier Medchemexpress LLC Supplier No. HYD033210MG
Disperse Orange 1 is an orange azo disperse dye (C18H14N4O2) used primarily for dyeing synthetic fibers such as polyester and nylon. Supplied in small laboratory pack sizes for research and analytical use, it is commonly used in textile dyeing studies and fluorescence applications. Accompanying datasheet and safety data sheet (SDS) provide handling and storage information.
  • High purity (approximately 96.6%) for analytical applications.
  • Molecular weight 318.33 for formulation calculations.
  • Suitable for dyeing polyester, nylon, and acrylic fibers.
  • Available in small laboratory pack sizes, including 10 mg, for method development.
  • Provided with a datasheet and SDS for safe handling and storage.