Learn More
Apexbio Technology LLC Disperse Red 1 2872-52-8 250mg
SDP
Catalog No. 5000093650
Encompass_Preferred
Disperse Red 1 (CAS 2872-52-8) is an azobenzene derivative employed as a photosensitive molecular tool It modulates biomolecular or polymer conformations via reversible photoinduced cis-trans isomerization upon exposure to specific wavelengths of light enabling optical regulation of cellular processes Disperse Red 1 exerts its biological effects primarily through light-triggered isomerization of its azo bond facilitating structural changes in target systems In experimental studies it is utilized to investigate cellular signaling pathways receptor activation mechanisms photocontrolled drug delivery and to regulate neuronal activity or membrane permeability in neurobiology and cell biology research Based on these properties Disperse Red 1 holds research potential in the photoregulation of biological processes optogenetics and experimental pharmacology
Catalog No. 50-000-93650 Supplier Apexbio Technology LLC Supplier No. A4617250
Disperse Red 1 (CAS 2872-52-8) is an azobenzene derivative employed as a photosensitive molecular tool It modulates biomolecular or polymer conformations via reversible photoinduced cis-trans isomerization upon exposure to specific wavelengths of light enabling optical regulation of cellular processes Disperse Red 1 exerts its biological effects primarily through light-triggered isomerization of its azo bond facilitating structural changes in target systems In experimental studies it is utilized to investigate cellular signaling pathways receptor activation mechanisms photocontrolled drug delivery and to regulate neuronal activity or membrane permeability in neurobiology and cell biology research Based on these properties Disperse Red 1 holds research potential in the photoregulation of biological processes optogenetics and experimental pharmacology