Learn More
Medchemexpress LLC Disperse Red 1 | 2872-52-8 | 95.0% | 1 ML
SDP
Catalog No. 5000410866
Encompass_Preferred
Disperse Red 1 is an azobenzene derivative and an azo textile dye, extensively used for dyeing polyester fabrics in the textile industry. This product is intended for research use only.
  • Designed for high-speed applications
  • Causes testicular toxicity in mice at certain concentrations
  • Increases frequency of micronuclei and causes mutations in human lymphocytes and HepG2 cell lines
  • Induces cytotoxic and genotoxic effects in mouse germ cells
Catalog No. 50-004-10866 Supplier Medchemexpress LLC Supplier No. HYD034210MM/1ML
Disperse Red 1 is an azobenzene derivative and an azo textile dye, extensively used for dyeing polyester fabrics in the textile industry. This product is intended for research use only.
  • Designed for high-speed applications
  • Causes testicular toxicity in mice at certain concentrations
  • Increases frequency of micronuclei and causes mutations in human lymphocytes and HepG2 cell lines
  • Induces cytotoxic and genotoxic effects in mouse germ cells