Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Disperse Red 13

Catalog No. 3648275G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-482-75G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-482-75G Supplier Sigma Aldrich Supplier No. 3648275G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Disperse Red 13

Specifications

CAS 3180-81-2
Melting Point 122°C to 129°C
Molecular Formula C16H17ClN4O3
Linear Formula ClC6H3(NO2)N=NC6H4N(C2H5)CH2CH2OH
Quantity 5 g
MDL Number MFCD00007208
Synonym 2-[4-(2-Chloro-4-nitrophenylazo)-N-ethylphenylamino]ethanol
Molecular Weight (g/mol) 348.78
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.