Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disperse Red 17, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013913 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3913 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3913 Supplier MilliporeSigma Supelco Supplier No. 7989225MG
Add to Cart
Add to Cart

Disperse red 17 is a dye, which is widely used in textile industry.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 3179-89-3
Physical Form Neat
Linear Formula C17H20N4O4
MDL Number MFCD00071927
Synonym 3-Methyl-4-(4 ′-nitrophenylazo)-N,N-bis(2-hydroxyethyl)aniline
Type Analytical Standard
Quantity 25 mg
Formula Weight 344.37
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.