Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disperse Yellow 9, ≥96.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015006 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5006 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5006 Supplier MilliporeSigma Supelco Supplier No. 3846425MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 6373-73-5
Physical Form Neat
Linear Formula (O2 N)2C6H3 NHC6H4 NH2
MDL Number MFCD00007851
Melting Point 187°C to 190°C
Synonym N-(2,4-Dinitrophenyl)-1,4-phenylenediamine; N-(2,4-Dinitrophenyl)-p-phenylenediamine
Type Analytical Standard
Quantity 25 mg
Formula Weight 274.23
Percent Purity ≥96.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.