Learn More
VELP Scientifica™ Dispersing Tool
SDP
Catalog No. 08418528
Change view
Click to view available options
Description:
Dispersing Tool
Sample Volume (Metric):
50 to 2000 mL
Catalog No. Description Sample Volume (Metric)
08-418-528 Dispersing Tool 50 to 2000 mL
1 options
Catalog No. 08-418-528 Supplier VELP Scientifica™ Supplier No. A00000464
Add to Cart
Add to Cart
Dispersing tool, suitable for emulsions and suspensions that require high final fineness.
SAMPLES AND APPLICATIONS
  • Preparation of foamy mixtures (ice cream, butter, margarine, icing) and spreadable creams
  • Preparation of tablets coatings
  • Preparation of pharmaceutical ointments and creams
  • Preparation of cosmetic products (lip balm, soap, shampoo, conditioner, creams, lipstick, mascara)
  • Preparation of pigments, inks, paints
  • Preparation of biodiesel and dispersion of petroleum derivatives (e.g., asphalt)
Specifications
Product Type Dispersing Tool
Sample Volume (Metric) 50 to 2000 mL
Immersion Length 40 to 175 mm
Autoclavable Autoclavable
Blade Material Stainless Steel
Material Stainless Steel
Description Dispersing Tool
Packaging Quantity 1 Ea.