Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caframo™ Dispersion Blade
GREENER_CHOICE

Catalog No. 01549748 Shop All Caframo Products
Change view
Click to view available options
Diameter (English):
3 in.
4 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English)
01-549-748 3 in.
01-549-747 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-549-748 Supplier Caframo™ Supplier No. A431
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dispersion Blade only with set screws, electropolished. For high shear mixing at medium to high speeds.

  • Electropolished 316 stainless steel
  • High shear
  • Broken particles are dispersed by the radial flow set up by the blade

Specifications

Diameter (English) 3 in.
Diameter (Metric) 75 mm
Type Dispersion Blade
Material Electropolished Stainless Steel (316)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.