Learn More
Caframo™ Dispersion Blade Set
GREENER_CHOICE
Catalog No. 01549749 Shop All Caframo Products
Change view
Click to view available options
Type:
Dispersion Blade
Catalog No. Type
01-549-749 Dispersion Blade
1 options
Catalog No. 01-549-749 Supplier Caframo™ Supplier No. A400SET
Add to Cart
Add to Cart
Quickly scale up your high shear mixing with this convenient set of three dispersion blades.
  • Set includes 48 mm (1 7/8 in.) A164, 75 mm (3 in.) A431, and 100 mm (4 in.) A441 blades with set screws
  • Blades are composed of electropolished 316 stainless steel with an 8 mm (5/16 in.) bore
  • Compatible with all Caframo shafts
  • To be used with BDC1850 or BDC3030 overhead stirrers
Specifications
Type Dispersion Blade
Material Electropolished Stainless Steel (316)