Learn More
Anton Paar Dispersion unit with integrated ultrasonication for liquid measurements in water to disperse particles in liquid medium (up to 2500 µm).
Catalog No. 5000580493
Encompass_Preferred
LIQUID FLOW DISPERSION UNITDispersion unit with integrated ultrasonication for liquid measurements inwater to disperse particles in liquid medium (up to 2500 µm).Flexible tank filling volume: 150-600 mL.Automated filling, rinsing, and draining.Minimum requirements to run:- outflow of the liquid (wastewater disposal connection or a tank)Recommended requirements to run:- water supply 0.1 - 3 bar
Catalog No. 50-005-80493 Supplier Anton Paar Supplier No. 261786
Please contact your sales representative for a quote or further ordering instructions.
LIQUID FLOW DISPERSION UNITDispersion unit with integrated ultrasonication for liquid measurements inwater to disperse particles in liquid medium (up to 2500 µm).Flexible tank filling volume: 150-600 mL.Automated filling, rinsing, and draining.Minimum requirements to run:- outflow of the liquid (wastewater disposal connection or a tank)Recommended requirements to run:- water supply 0.1 - 3 bar