Learn More
Enterprise Technology Solutions BELKIN F2CD032b DisplayPort Male to Female VGA Adapter, Black
SDP
Catalog No. 502096746
Encompass_Preferred
Belkin Displayport to VGA adapter supports PC resolutions of up to 1920x1200 and HDTV resolutions up to 1080p. This high-resolution adapter offers plug and play functionality to provide an active digital to analog conversion without software or drivers. F2CM032b is a video only adapter; please check your monitor/projector/display settings to your desired graphic resolution.
Catalog No. 50-209-6746 Supplier Enterprise Technology Solutions Supplier No. F2CD032B
Belkin Displayport to VGA adapter supports PC resolutions of up to 1920x1200 and HDTV resolutions up to 1080p. This high-resolution adapter offers plug and play functionality to provide an active digital to analog conversion without software or drivers. F2CM032b is a video only adapter; please check your monitor/projector/display settings to your desired graphic resolution.