Learn More
HF Scientific Inc Free Chlorine DPD Reagent Dispenser, 400 Tests (2x200), 5ml Sample Size For HF scientific and Hach Photometers
Catalog No. NC1849571
Encompass_Preferred
This HF scientific portable dispenser accurately delivers the correct volume of DPD reagent for free chlorine. It is supplied with enough DPD powder for 400 tests. To use, simply invert the dispenser over the sample and press the button to dispense the reagent. The powder dissolves quickly in the sample, leaving no messy residue behind. It is a 5ml sample size, with formulations for use with HF scientific and Hach photometers.
Catalog No. NC1849571 Supplier HF Scientific Inc Supplier No. 10445A
This HF scientific portable dispenser accurately delivers the correct volume of DPD reagent for free chlorine. It is supplied with enough DPD powder for 400 tests. To use, simply invert the dispenser over the sample and press the button to dispense the reagent. The powder dissolves quickly in the sample, leaving no messy residue behind. It is a 5ml sample size, with formulations for use with HF scientific and Hach photometers.