Learn More
Electron Microscopy Sciences DispoCut™ Dissecting Board, 12 x 19 in
SDP
Catalog No. 501900434
Encompass_Preferred
Strong and inexpensive disposable cutting board provides a clean, safer and more efficient way of handling infectious tissue specimens. Printed with helpful imperial and metric dimensional scales in inches, centimeters and millimeters on both sides. Made using a copolymer resin that increases impact and high and low temperature performance and flexibility.
Chemically inert. At regular temperatures most oils, solvents, and water have no effect, allowing it to perform under adverse weather conditions or as a product component exposed to harsh chemicals. Surface will not dull knifes. Perfect for use with dissecting pins.
May be wiped down with alcohol with no effect on the material. Ethylene oxide gas sterilization has no effect of an kind on DispoCut and may be repeated any number of times.
Size: 12 x 19 in
Catalog No. 50-190-0434 Supplier Electron Microscopy Sciences Supplier No. 6330840
Strong and inexpensive disposable cutting board provides a clean, safer and more efficient way of handling infectious tissue specimens. Printed with helpful imperial and metric dimensional scales in inches, centimeters and millimeters on both sides. Made using a copolymer resin that increases impact and high and low temperature performance and flexibility.
Chemically inert. At regular temperatures most oils, solvents, and water have no effect, allowing it to perform under adverse weather conditions or as a product component exposed to harsh chemicals. Surface will not dull knifes. Perfect for use with dissecting pins.
May be wiped down with alcohol with no effect on the material. Ethylene oxide gas sterilization has no effect of an kind on DispoCut and may be repeated any number of times.
Size: 12 x 19 in