Learn More
Thomas Scientific DispoCutT Dissecting Board, 9 x 12 in. (229 x 305 mm)
SDP
Catalog No. 501461244
Encompass
DispoCut™ Dissecting Board, 9 x 12 in. (229 x 305 mm) 12/pk, 48/case Strong yet inexpensive enough to throw away Provides a clean, safe and more efficient way of handling infectious tissue specimens Can be used on both sides — a great money saving feature. Printed with helpful imperial and metric dimensional scales in inches, centimeters and millimeters. Sterilization: DispoCut™ may be wiped down with alcohol with no effect on the material. Temperature performance range: –27 ÝC to 71 ÝC (17 ÝF to 160 ÝF) Melting point: 162 ÝC (324 ÝF)
Catalog No. 50-146-1244 Supplier Thomas Scientific Supplier No. 1159N31
DispoCut™ Dissecting Board, 9 x 12 in. (229 x 305 mm) 12/pk, 48/case Strong yet inexpensive enough to throw away Provides a clean, safe and more efficient way of handling infectious tissue specimens Can be used on both sides — a great money saving feature. Printed with helpful imperial and metric dimensional scales in inches, centimeters and millimeters. Sterilization: DispoCut™ may be wiped down with alcohol with no effect on the material. Temperature performance range: –27 ÝC to 71 ÝC (17 ÝF to 160 ÝF) Melting point: 162 ÝC (324 ÝF)