Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ Disposable 100mL Reagent Reservoirs
GSA_VAGREENER_CHOICE

Catalog No. 12111084 Shop All OHAUS Products
Change view
Click to view available options
Bottom Shape:
Flat
V-bottom
Quantity:
100/Cs.
20/Cs.
200/Cs.
50/Cs.
Packaging:
1 reservoir per bag
5 reservoirs per bag
Bulk
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bottom Shape Quantity Packaging
12-111-084 Flat 100/Cs. Bulk
12-111-082 V-bottom 50/Cs. 5 reservoirs per bag
12-111-083 V-bottom 20/Cs. 1 reservoir per bag
12-111-098 Flat 200/Cs. 5 reservoirs per bag
12-111-100 V-bottom 100/Cs. Bulk
12-111-099 V-bottom 100/Cs. 1 reservoir per bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 12-111-084 Supplier BIOTIX™ Supplier No. 63300119
Get Extra Pipette Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use with multi-channel pipetting applications and can accommodate up to 12-channel pipettes.

  • Angled sides and a V-shaped bottom allow maximum recovery of reagents
  • All reservoirs feature internal graduated volume lines and pour-spouts on the corners
  • Made from polystyrene

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Description With angled sides and V-shaped bottom
Capacity (Metric) 100 mL
Material Polystyrene
Sterility Non-sterile
Type Reservoir
For Use With (Application) Filling Multi-channel Pipettors
Packaging Bulk
Quantity 100/Cs.
Autoclavable Autoclavable
Channels 12
Disposable Single-use
Bottom Shape Flat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.