Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valutek™ Disposable 5 Snap Labcoats

Catalog No. 191500838
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-0838 X-Large
19-150-0841 4X-Large
19-150-0837 Large
19-150-0836 Medium
19-150-0839 2X-Large
19-150-0835 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-150-0838 Supplier Valutek™ Supplier No. VTLBCTXL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Product Type Controlled Environments Lab Coat
Size X-Large
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
No. of Pockets 3
Certifications/Compliance ISO 7
For Use With (Application) Used in electronic medical device and pharmaceuticals industries
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.