Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Disposable Alcohol Swabs
SCI_ED

Catalog No. S17032 Shop All BD Diagnostic Systems Products
This item is not returnable. View return policy
Catalog No. S17032 Supplier BD Supplier No. 326895

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Easy to open and ready to use.

Be sure you always have a good supply of double-thick alcohol pads on hand for blood collecting. Impregnated with 13mL of isopropyl alcohol, these 1.2 × 1.6 in. (3 × 4cm) pads come individually sealed in foil envelopes.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Alcohol Swab
Composition 70% Isopropyl Alcohol
Disposable Single-use
For Use With (Application) Household applications
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.