Learn More
Sigma Aldrich Fine Chemicals Biosciences Disposable aluminum dishes
Catalog No. 502901857
Encompass
Smooth bottom. Use for weighing dispensing evaporation or as a dust cover. Temp. limit 1 250f (676C).
Catalog No. 50-290-1857 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1548571PAK
Smooth bottom. Use for weighing dispensing evaporation or as a dust cover. Temp. limit 1 250f (676C).