Learn More
Med Vet International Disposable Anesthesia Mask, Vertical ID, J0298DC, 1.75in x 2.25in, Each
Catalog No. 5000583091
Encompass_Preferred
A crystal clear plastic material with inflation/deflation valve for better patient fitting. Inexpensive. 15mm OD fitting. Five different sizes. This one-way valve inflates with any standard syringe.
Catalog No. 50-005-83091 Supplier Med Vet International Supplier No. JOR298DC
A crystal clear plastic material with inflation/deflation valve for better patient fitting. Inexpensive. 15mm OD fitting. Five different sizes. This one-way valve inflates with any standard syringe.