Learn More
Electron Microscopy Sciences Disposable Base Mold, 7 x 7 mm, 5 mm height
SDP
Catalog No. 5025510
Encompass_Preferred
Our Disposable Base Mold is designed to fit all existing cassettes. Because our base molds are disposable you save time on cleaning and storing and they offer you less exposure to hazardous chemicals.
Size: 7 x 7 mm
Height: 5 mm
Catalog No. 50-255-10 Supplier Electron Microscopy Sciences Supplier No. 6235207
Our Disposable Base Mold is designed to fit all existing cassettes. Because our base molds are disposable you save time on cleaning and storing and they offer you less exposure to hazardous chemicals.
Size: 7 x 7 mm
Height: 5 mm