Learn More
Ted Pella Inc Disposable Base Molds, 24 x 24 x 5mm, 500/box
SDP
Catalog No. NC9348428
Encompass
Disposable base molds offer ease and convenience of operation. They are inexpensive enough to be discarded after use, yet strong enough to be reused. Made of PVC, they offer excellent thermal exchange. They have a smooth interior finish and rounded corners to facilitate their removal. Also, they are available in the same variety of sizes as metal molds and can be used with the same styles or types of cassettes and embedding rings.
Catalog No. NC9348428 Supplier Ted Pella Inc Supplier No. 271473
Disposable base molds offer ease and convenience of operation. They are inexpensive enough to be discarded after use, yet strong enough to be reused. Made of PVC, they offer excellent thermal exchange. They have a smooth interior finish and rounded corners to facilitate their removal. Also, they are available in the same variety of sizes as metal molds and can be used with the same styles or types of cassettes and embedding rings.