Learn More
Alkali Scientific Disposable Base Molds for Histology & Tissue Embedding, 500/Case, 25x32x5mm
SDP
Catalog No. 5000662385
Encompass_Preferred
Disposable Base Molds are designed for reliable and consistent paraffin embedding in histology and pathology laboratories. Manufactured from high quality plastic, these molds feature uniform wall thickness and precise dimensions that help maintain optimal tissue orientation and reproducible paraffin block formation during embedding procedures. The smooth interior surface allows easy paraffin release, minimizing block distortion and reducing the risk of tissue damage during removal. Disposable construction eliminates cleaning and reuse, helping laboratories improve workflow efficiency while reducing the potential for cross contamination. Compatible with standard embedding rings and tissue cassettes, these base molds support routine histology, pathology diagnostics, biomedical research, and educational laboratory applications.
Catalog No. 50-006-62385 Supplier Alkali Scientific Supplier No. DBM04
Disposable Base Molds are designed for reliable and consistent paraffin embedding in histology and pathology laboratories. Manufactured from high quality plastic, these molds feature uniform wall thickness and precise dimensions that help maintain optimal tissue orientation and reproducible paraffin block formation during embedding procedures. The smooth interior surface allows easy paraffin release, minimizing block distortion and reducing the risk of tissue damage during removal. Disposable construction eliminates cleaning and reuse, helping laboratories improve workflow efficiency while reducing the potential for cross contamination. Compatible with standard embedding rings and tissue cassettes, these base molds support routine histology, pathology diagnostics, biomedical research, and educational laboratory applications.