Learn More
Cole-Parmer Sterile Multichannel Disposable Solutions Basin
Supplier:  Cole-Parmer 2481035
Encompass_Preferred
Constructed of white polystyrene for easy side viewing. Angled sides and V-shaped bottoms allow efficient and nearly complete sample withdrawal. Basins hold up to 55mL, and are available for non-sterile and sterile applications.
Catalog No. NC1495398