Learn More
Cole-Parmer Cole-Parmer Polystyrene (PS) Four-Spout Beaker, 10 mL; 1000/Pk
Catalog No. NC0158122 Shop All Cole-Parmer Products
Encompass_Preferred
These durable polystyrene beakers feature four-way pout spouts for easy pouring. Beakers can withstand temperatures to 180°F (82°C).
Catalog No. NC0158122 Supplier Cole-Parmer Supplier No. 0600555
Add to Cart
Add to Cart
These durable polystyrene beakers feature four-way pout spouts for easy pouring. Beakers can withstand temperatures to 180°F (82°C).