Learn More
Sigma Aldrich Fine Chemicals Biosciences Disposable beakers capacit
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2453991PAK
Encompass
Translucent PP with three dripless pouring spoutsMolded graduationsflat bottoms keep magnetic stir bars centeredAutoclavable
Catalog No. 50-290-1604