Learn More
Sigma Aldrich Fine Chemicals Biosciences Disposable beakers capacit
Catalog No. 502900407
Encompass
Translucent PP with three dripless pouring spoutsMolded graduationsflat bottoms keep magnetic stir bars centeredAutoclavable
Catalog No. 50-290-0407 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2454291PAK
Translucent PP with three dripless pouring spoutsMolded graduationsflat bottoms keep magnetic stir bars centeredAutoclavable