Learn More
Electron Microscopy Sciences Disposable Bibulous Paper Book 10
SDP
Catalog No. 5033479
Encompass_Preferred
Highly absorbent white fiber disposable paper. Recommended for drying microscope slides and other small lab glass items. Lint and dust free.
Catalog No. 50-334-79 Supplier Electron Microscopy Sciences Supplier No. 7008610
Highly absorbent white fiber disposable paper. Recommended for drying microscope slides and other small lab glass items. Lint and dust free.