Learn More
World Precision Instrument Disposable Biopsy Punch, Various Sizes-501818
SDP
Catalog No. 502521400
Encompass
Our disposable biopsy punches have a seamless, stainless steel, razor sharp cutting edge. They are individually packaged in color-coded, transparent, sturdy blister pack to protect the cutting edge. Its ribbed handle provides maximum control and comfort.
Catalog No. 50-252-1400 Supplier World Precision Instrument Supplier No. 501818
Our disposable biopsy punches have a seamless, stainless steel, razor sharp cutting edge. They are individually packaged in color-coded, transparent, sturdy blister pack to protect the cutting edge. Its ribbed handle provides maximum control and comfort.