Learn More
Epredia™ Disposable Blade Carriers
Catalog No. p-4529865 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Blade Carrier E
For Use With (Equipment):
HM 310, HM 315 and HM 325
Catalog No. Product Type For Use With (Equipment)
70-580-0 Blade Carrier E HM 310, HM 315 and HM 325
1 options
Catalog No. 70-580-0 Supplier Epredia™ Supplier No. 705800
Only null left
Add to Cart
Add to Cart
Utilize low- and high-profile blades with the quick clamp system of Epredia™ Disposable Blade Carriers.
Disposable Blade Carriers
  • Blade angle and blade adjustment levers are removable to help protect user settings (Version 2 and 3 units only)
  • Concentric lever locks onto the dovetail to help reduce accidental movement of the holder (Version 2 and 3 units only)
  • Quick clamp system for high- and low-profile blades
  • Adjustable clearance angle
  • Moveable knife guard covers the blade entirely in any position
  • Knife carrier base included
Specifications
Description Blade carrier E
Product Type Blade Carrier E
For Use With (Equipment) HM 310, HM 315 and HM 325