Learn More
StatLab Disposable Blade Remover
SDP
Catalog No. 21033273
Change view
Click to view available options
Quantity:
5 Pk.
Catalog No. Quantity
21-033-273 5 Pk.
1 options
Catalog No. 21-033-273 Supplier StatLab Supplier No. BLR00201P
Convenient method for removal of scalpel blades
  • 5 per pack
  • Insert blade and handle into the slot, press and pull. Blade falls safely into a compartment that stores up to 100 blades. Sold individually.
Specifications
Product Type Disposable Blade Remover
For Use With (Equipment) Specimens
Quantity 5 Pk.