Learn More
Med Vet International Disposable Blood Pressure Cuff, White, 3.5cm, J0563Q, 5 Pack
Catalog No. 5000583822
Encompass_Preferred
These cuffs are made from a soft white vinyl. They feature a Velcro closure and will fit all models of blood pressure measurement systems. A single tube. Made in the USA, and inexpensive. White. 5/pk.Proper Size is 40 percent of the circumference of the limb when the cuff is being applied These sizes should cover just about all small animals. Cuff size ranges in centimeters.
Catalog No. 50-005-83822 Supplier Med Vet International Supplier No. JOR563Q
These cuffs are made from a soft white vinyl. They feature a Velcro closure and will fit all models of blood pressure measurement systems. A single tube. Made in the USA, and inexpensive. White. 5/pk.Proper Size is 40 percent of the circumference of the limb when the cuff is being applied These sizes should cover just about all small animals. Cuff size ranges in centimeters.