Learn More
Med Vet International Disposable Blood Pressure Cuffs, 1.5cm, J0563X, White, 5 Pack
Catalog No. 5000583811
Encompass_Preferred
1.5cm W 4.3-6.4 L (#1) These cuffs are made from a soft white vinyl. They feature a Velcro closure and will fit all models of blood pressure measurement systems. A single tube. Made in the USA, and inexpensive. White. 5/pk. Proper Size is 40 percent of th
Catalog No. 50-005-83811 Supplier Med Vet International Supplier No. JOR563X
1.5cm W 4.3-6.4 L (#1) These cuffs are made from a soft white vinyl. They feature a Velcro closure and will fit all models of blood pressure measurement systems. A single tube. Made in the USA, and inexpensive. White. 5/pk. Proper Size is 40 percent of th