Learn More
RWD LIFE SCIENCE INC Disposable Cell Counting Slide DS-50,2 sample/pcs,50 pcs/Package, Capacity 10 microliter
SDP
Catalog No. NC3404392
Encompass_Preferred
Disposable cell counting slide consisting of two independent counting cells, is used in automatic cell counter C100/C100-SE/ C100-PRO. It has good compatibility and can be adapted to the same type of image automatic cell counterspecifications: 2 sample/pcs,50 pcs/Package; Capacity: 10 microliter
Catalog No. NC3404392 Supplier RWD LIFE SCIENCE INC Supplier No. DS50
Disposable cell counting slide consisting of two independent counting cells, is used in automatic cell counter C100/C100-SE/ C100-PRO. It has good compatibility and can be adapted to the same type of image automatic cell counterspecifications: 2 sample/pcs,50 pcs/Package; Capacity: 10 microliter