Learn More
Research Products International Corp Disposable Charcoal Cartridges, 3 per Package
SDP
Catalog No. 50550317
Encompass
Disposable Charcoal Cartridges absorb volatile 35S released during metabolic labeling procedures inside CO2 incubators. Petri dish cartridges contain activated charcoal sealed in a highly porous filter membrane. As CO2 flows over the cartridge, volatile 35S becomes absorbed by the charcoal. Following use, cartridges can be discarded as radioactive waste.
WARNING  This product can expose you to chemicals including Carbon Black, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-550-317 Supplier Research Products International Corp Supplier No. 248849
Disposable Charcoal Cartridges absorb volatile 35S released during metabolic labeling procedures inside CO2 incubators. Petri dish cartridges contain activated charcoal sealed in a highly porous filter membrane. As CO2 flows over the cartridge, volatile 35S becomes absorbed by the charcoal. Following use, cartridges can be discarded as radioactive waste.
WARNING  This product can expose you to chemicals including Carbon Black, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.